Anti-FOXL1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9211CEA 448 CHF
ABGEAP9211C
Anti-FOXL1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FOXL1 (forkhead box L1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR