Anti-ALOX15B Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9200BEA 407 CHF
ABGEAP9200B
Anti-ALOX15B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ALOX15B (arachidonate 15-lipoxygenase, type B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR