Anti-GSTP1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9199CEA 448 CHF
ABGEAP9199C
Anti-GSTP1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GSTP1 (glutathione S-transferase pi 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR