Anti-AFMID Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9175AEA 405 CHF
ABGEAP9175A
Anti-AFMID Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AFMID (arylformamidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR