Anti-HMCES Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9160AEA 378 CHF
ABGEAP9160A
Anti-HMCES Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HMCES (5-hydroxymethylcytosine (hmC) binding, ES cell-specific)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR