Anti-UTY Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9074BEA 405 CHF
ABGEAP9074B
Anti-UTY Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UTY (ubiquitously transcribed tetratricopeptide repeat containing, Y-linked)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR