Anti-KDR Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9054AEA 407 CHF
ABGEAP9054A
Anti-KDR Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KDR (kinase insert domain receptor (a type III receptor tyrosine kinase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR