Anti-APP Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9053AEA 378 CHF
ABGEAP9053A
Anti-APP Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR