Anti-EXTL3 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9048AEA 407 CHF
ABGEAP9048A
Anti-EXTL3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: EXTL3 (exostosin-like glycosyltransferase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR