Anti-SFRP1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9037AEA 405 CHF
ABGEAP9037A
Anti-SFRP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SFRP1 (secreted frizzled-related protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR