Anti-CLASP2 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9035AEA 378 CHF
ABGEAP9035A
Anti-CLASP2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CLASP2 (cytoplasmic linker associated protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR