Anti-CDK5RAP1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9034BEA 405 CHF
ABGEAP9034B
Anti-CDK5RAP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CDK5RAP1 (CDK5 regulatory subunit associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR