Anti-STK40 Rabbit Polyclonal Antibody [clone: 345CT19.6.2]

Supplier: Abgent
ABGEAP9026BEA 407 CHF
ABGEAP9026B
Anti-STK40 Rabbit Polyclonal Antibody [clone: 345CT19.6.2]
Antibodies
Type: Primary
Antigen: STK40 (serine/threonine kinase 40)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR