Anti-TBC1D32 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9019AEA 378 CHF
ABGEAP9019A
Anti-TBC1D32 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TBC1D32 (TBC1 domain family, member 32)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR