Anti-EXOC5 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9006BEA 407 CHF
ABGEAP9006B
Anti-EXOC5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: EXOC5 (exocyst complex component 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR