Anti-EDIL3 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9005CEA 378 CHF
ABGEAP9005C
Anti-EDIL3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: EDIL3 (EGF-like repeats and discoidin I-like domains 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR