Anti-E2F5 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9003AEA 405 CHF
ABGEAP9003A
Anti-E2F5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: E2F5 (E2F transcription factor 5, p130-binding)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR