Anti-DPP10 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9000CEA 407 CHF
ABGEAP9000C
Anti-DPP10 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DPP10 (dipeptidyl-peptidase 10 (non-functional))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR