Anti-FCGR2C Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8992BEA 405 CHF
ABGEAP8992B
Anti-FCGR2C Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FCGR2C (Fc fragment of IgG, low affinity IIc, receptor for (CD32) (gene/pseudogene))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR