Anti-IL36B Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8956AEA 407 CHF
ABGEAP8956A
Anti-IL36B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL36B (interleukin 36, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR