Anti-TGOLN2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8955BEA 405 CHF
ABGEAP8955B
Anti-TGOLN2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TGOLN2 (trans-golgi network protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR