Anti-CDC5L Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8949CEA 419 CHF
ABGEAP8949C
Anti-CDC5L Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CDC5L (cell division cycle 5-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR