Anti-SLC8A1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8939CEA 405 CHF
ABGEAP8939C
Anti-SLC8A1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLC8A1 (solute carrier family 8 (sodium/calcium exchanger), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR