Anti-ALDH18A1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8938CEA 407 CHF
ABGEAP8938C
Anti-ALDH18A1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ALDH18A1 (aldehyde dehydrogenase 18 family, member A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR