Anti-LMX1B Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8935CEA 405 CHF
ABGEAP8935C
Anti-LMX1B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LMX1B (LIM homeobox transcription factor 1, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR