Anti-ETV6 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8917AEA 419 CHF
ABGEAP8917A
Anti-ETV6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ETV6 (ets variant 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR