Anti-KIR2DL5B Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8909CEA 405 CHF
ABGEAP8909C
Anti-KIR2DL5B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KIR2DL5B (killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 5B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR