Anti-ADAMTS18 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8906BEA 405 CHF
ABGEAP8906B
Anti-ADAMTS18 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ADAMTS18 (ADAM metallopeptidase with thrombospondin type 1 motif, 18)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR