Anti-ALG14 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8903CEA 378 CHF
ABGEAP8903C
Anti-ALG14 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ALG14 (ALG14, UDP-N-acetylglucosaminyltransferase subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR