Anti-INPP4B Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8902CEA 405 CHF
ABGEAP8902C
Anti-INPP4B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: INPP4B (inositol polyphosphate-4-phosphatase, type II, 105kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR