Anti-TNFSF11 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8895AEA 378 CHF
ABGEAP8895A
Anti-TNFSF11 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TNFSF11 (tumor necrosis factor (ligand) superfamily, member 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR