Anti-B4GALT1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8892BEA 378 CHF
ABGEAP8892B
Anti-B4GALT1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: B4GALT1 (UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR