Anti-PTGER2 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8862CEA 378 CHF
ABGEAP8862C
Anti-PTGER2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PTGER2 (prostaglandin E receptor 2 (subtype EP2), 53kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR