Anti-ADCY8 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8858CEA 407 CHF
ABGEAP8858C
Anti-ADCY8 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ADCY8 (adenylate cyclase 8 (brain))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR