Anti-CASD1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8857AEA 407 CHF
ABGEAP8857A
Anti-CASD1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CASD1 (CAS1 domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR