Anti-KLF16 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8855CEA 407 CHF
ABGEAP8855C
Anti-KLF16 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KLF16 (Kruppel-like factor 16)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR