Anti-PCCB Rabbit Polyclonal Antibody [clone: 399CT9.3.4]

Supplier: Abgent
ABGEAP8843CEA 407 CHF
ABGEAP8843C
Anti-PCCB Rabbit Polyclonal Antibody [clone: 399CT9.3.4]
Antibodies
Type: Primary
Antigen: PCCB (propionyl CoA carboxylase, beta polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR