Anti-UBAP1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8828AEA 407 CHF
ABGEAP8828A
Anti-UBAP1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: UBAP1 (ubiquitin associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR