Anti-CANT1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8822AEA 407 CHF
ABGEAP8822A
Anti-CANT1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CANT1 (calcium activated nucleotidase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR