Anti-ASPSCR1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8821BEA 448 CHF
ABGEAP8821B
Anti-ASPSCR1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ASPSCR1 (alveolar soft part sarcoma chromosome region, candidate 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR