Anti-IL18RAP Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8797AEA 448 CHF
ABGEAP8797A
Anti-IL18RAP Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL18RAP (interleukin 18 receptor accessory protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR