Anti-IL12A Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8796BEA 448 CHF
ABGEAP8796B
Anti-IL12A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL12A (interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR