Anti-RASL11B Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8789AEA 407 CHF
ABGEAP8789A
Anti-RASL11B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RASL11B (RAS-like, family 11, member B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR