Anti-CYP8B1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8787BEA 405 CHF
ABGEAP8787B
Anti-CYP8B1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CYP8B1 (cytochrome P450, family 8, subfamily B, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR