Anti-IL10RA Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8768CEA 378 CHF
ABGEAP8768C
Anti-IL10RA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL10RA (interleukin 10 receptor, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR