Anti-BMI1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8756AEA 405 CHF
ABGEAP8756A
Anti-BMI1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BMI1 (BMI1 polycomb ring finger oncogene)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR