Anti-DDX39B Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8740BEA 419 CHF
ABGEAP8740B
Anti-DDX39B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DDX39B (DEAD (Asp-Glu-Ala-Asp) box polypeptide 39B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR