Anti-DERL2 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8707BEA 407 CHF
ABGEAP8707B
Anti-DERL2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DERL2 (derlin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR