Anti-CACNA2D1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8699AEA 407 CHF
ABGEAP8699A
Anti-CACNA2D1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CACNA2D1 (calcium channel, voltage-dependent, alpha 2/delta subunit 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR