Anti-LTF Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP8698AEA 407 CHF
ABGEAP8698A
Anti-LTF Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LTF (lactotransferrin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR